Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ABS 047-14-02 binds free canine NGAL.
This antibody shows no cross-reaction with human, mouse ,or rat.
ABS 047-14-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibody ABS 047-17-02, giving a detection limit of 5 pg/mL in an unoptimized buffer assay.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
NGAL (Neutrophil gelatinase-associated lipocalin, lipocalin 2, siderocalin and neutrophil lipocalin) is a 25 kDa glycosylated single chain monomer and member of the lipocalin family of proteins that bind and transport small lipophilic molecules. NGAL is released by activated neutrophils, can form dimers and small amounts of higher oligomers, and complexes with matrix metalloproteinase 9 (MMP-9, gelatinase B). Low level expression of NGAL in a variety of epithelia may be increased in inflammation or cancers. Expression of NGAL in the kidney is dramatically increased by ischemic or nephrotoxic kidney injury.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:6526; lipocalin 2 (oncogene 24p3); Lipocalin2; OTTHUMP00000022240; RP23-161B9.11; uterocalin
Entrez Gene ID: (Dog) 491320
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support